Anti-POU5F1 Mouse Monoclonal Antibody [clone: 1B11]
ABNOH00005460-M05
: Abnova
- Antibody type:Primary
- Antigen name:POU Class 5 Homeobox 1
- Antigen symbol:POU5F1
- Clonality:Monoclonal
- Clone:1B11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5460
- Antigen synonyms:POU domain class 5 transcription factor 1|Oct-4|OCT3|OTF3.|Octamer-binding transcription factor 3|Oct-3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: POU5F1
Clonality: Monoclonal
Clone: 1B11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human