Anti-SERPINI2 Mouse Polyclonal Antibody
ABNOH00005276-B01
: Abnova
- Antibody type:Primary
- Antigen name:Serpin peptidase inhibitor, clade I, member 2
- Antigen symbol:SERPINI2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5276
- Antigen synonyms:MEPI|TSA2004|PANCPIN|serpin peptidase inhibitor|PI14|protease inhibitor 14,|member 2|SERPINI2|serine (or cysteine) proteinase inhibitor|clade I (neuroserpin)|SERPIN12|myoepithelium-derived serine protease inhibitor|clade I (pancpin)
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Type: Primary
Antigen: SERPINI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human