Anti-SERPINB9 Mouse Monoclonal Antibody [clone: 1F5]
Catalog # ABNOH00005272-M06
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Serpin peptidase inhibitor, clade B (ovalbumin), member 9
- Antigen symbol:SERPINB9
- Clonality:Monoclonal
- Clone:1F5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:5272
- Antigen synonyms:CAP-3|CAP3|PI-9|PI9
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SERPINB9
Clonality: Monoclonal
Clone: 1F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human