Order Entry
Poland
ContactUsLinkComponent
Anti-PFKP Mouse Polyclonal Antibody
Anti-PFKP Mouse Polyclonal Antibody
Catalog # ABNOH00005214-B01
Supplier:  Abnova
undefined
Anti-PFKP Mouse Polyclonal Antibody
Catalog # ABNOH00005214-B01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Phosphofructokinase, platelet
  • Antigen symbol:
    PFKP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    5214
  • Antigen synonyms:
    Phosphofructo-1-kinase isozyme C|Phosphohexokinase|platelet type|6-phosphofructokinase|type C|Phosphofructokinase 1|PFK-C
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: PFKP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human