Anti-PAX6 Rabbit Polyclonal Antibody
Catalog # ABNOH00005080-D01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Paired box 6
- Antigen symbol:PAX6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5080
- Antigen synonyms:MGDA|Paired Box Protein-6|Oculorhombin|WAGR|Keratitis)|Keratitis|AN2|Aniridia|Aniridia Type II Protein|Paired Box Gene 6 (Aniridia
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: PAX6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse