Anti-REG3A Rabbit Polyclonal Antibody
Catalog # ABNOH00005068-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Regenerating islet-derived protein 3 alpha
- Antigen symbol:REG3A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:5068
- Antigen synonyms:PBCGF|HIP|INGAP|REG-III|PAP|regenerating family member 3 alpha|PAP1|HIP/PAP|PAP-H|REG3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: REG3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human