Order Entry
Poland
ContactUsLinkComponent
 
Anti-NEU2 Mouse Monoclonal Antibody [clone: 2E5]
  ABNOH00004759-M04
 :  Abnova
undefined
Anti-NEU2 Mouse Monoclonal Antibody [clone: 2E5]
  ABNOH00004759-M04
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Neuraminidase 2
  • Antigen symbol:
    NEU2
  • Clonality:
    Monoclonal
  • Clone:
    2E5
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    4759
  • Antigen synonyms:
    Sialidase 2|SIAL2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: NEU2
Clonality: Monoclonal
Clone: 2E5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human