Order Entry
Poland
ContactUsLinkComponent
Anti-MMP12 Rabbit Polyclonal Antibody
Anti-MMP12 Rabbit Polyclonal Antibody
Catalog # ABNOH00004321-D01P
Supplier:  Abnova
undefined
Anti-MMP12 Rabbit Polyclonal Antibody
Catalog # ABNOH00004321-D01P
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Matrix Metalloproteinase 12
  • Antigen symbol:
    MMP12
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    4321
  • Antigen synonyms:
    matrix metalloproteinase 12|MGC138506|macrophage metalloelastase|MME|HME|matrix metalloproteinase 12 (macrophage elastase)|macrophage elastase|MMP12|matrix metallopeptidase 12 (macrophage elastase)
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: MMP12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat