Anti-MEF2D Mouse Polyclonal Antibody
Catalog # ABNOH00004209-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Myocyte enhancer factor 2D
- Antigen symbol:MEF2D
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4209
- Antigen synonyms:polypeptide D (myocyte enhancer factor 2D)|myocyte enhancer factor 2D/deleted in azoospermia associated protein 1 fusion protein|myocyte-specific enhancer factor 2D|MADS box transcription enhancer factor 2|MEF2D/DAZAP1 fusion
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: MEF2D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human