Anti-MATK Mouse Monoclonal Antibody [clone: 2C5]
Catalog # ABNOH00004145-M05
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Megakaryocyte-associated Tyrosine Kinase
- Antigen symbol:MATK
- Clonality:Monoclonal
- Clone:2C5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4145
- Antigen synonyms:HYLTK|Tyrosine-protein kinase CTK|CTK|HHYLTK|Lsk|Protein kinase HYL|Megakaryocyte-associated tyrosine-protein kinase|CHK|Hematopoietic consensus tyrosine-lacking kinase|HYL
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: MATK
Clonality: Monoclonal
Clone: 2C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human