Anti-MAK Rabbit Polyclonal Antibody
Catalog # ABNOH00004117-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Serine/threonine-protein kinase MAK
- Antigen symbol:MAK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:4117
- Antigen synonyms:RP62|dJ417M14.2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MAK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse