Anti-KCNJ11 Mouse Polyclonal Antibody
Catalog # ABNOH00003767-A01
Dostawca: Abnova
Dane techniczne
- Antibody type:Primary
- Antigen name:ATP-sensitive inward rectifier potassium channel 11
- Antigen symbol:KCNJ11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3767
- Antigen synonyms:potassium channel inwardly rectifing subfamily J member 11|IKATP|inward rectifier K(+) channel Kir6.2|TNDM3|potassium channel|BIR|beta-cell inward rectifier subunit|HHF2|inwardly rectifying potassium channel KIR6.2|KIR6.2|ATP-sensitive inward rectifier potassium channel 11|inwardly|PHHI
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µl
- Pk:50 µl
Dane techniczne
Informacje o tym produkcie
Type: Primary
Antigen: KCNJ11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human