Anti-JAK3 Mouse Monoclonal Antibody [clone: 2A5]
Catalog # ABNOH00003718-M03
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Janus kinase 3
- Antigen symbol:JAK3
- Clonality:Monoclonal
- Clone:2A5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:3718
- Antigen synonyms:L-JAK|JAK-3|JAKL|LJAK|JAK3_HUMAN
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: JAK3
Clonality: Monoclonal
Clone: 2A5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human