Anti-ITGA6 Mouse Monoclonal Antibody [clone: 4C1]
Catalog # ABNOH00003655-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Integrin Alpha 6
- Antigen symbol:ITGA6
- Clonality:Monoclonal
- Clone:4C1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3655
- Antigen synonyms:CD49 antigen-like family member F|integrin alpha-6|CD49f|ITGA6|VLA-6|ITGA6B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ITGA6
Clonality: Monoclonal
Clone: 4C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human