Anti-PDX1 Mouse Monoclonal Antibody [clone: 2E4]
ABNOH00003651-M03
: Abnova
- Antibody type:Primary
- Antigen name:Pancreatic and duodenal homeobox 1
- Antigen symbol:PDX1
- Clonality:Monoclonal
- Clone:2E4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:3651
- Antigen synonyms:Islet/duodenum homeobox-1|IPF-1|Insulin promoter factor 1|Pancreas/duodenum homeobox protein 1|IPF1|Somatostatin-transactivating factor 1|Insulin upstream factor 1|IUF-1|PDX-1|GSF|STF-1|Glucose-sensitive factor|PDX1|IDX-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: PDX1
Clonality: Monoclonal
Clone: 2E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human