Anti-HSF4 Mouse Monoclonal Antibody [clone: 2B3]
Catalog # ABNOH00003299-M05
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Heat shock transcription factor 4
- Antigen symbol:HSF4
- Clonality:Monoclonal
- Clone:2B3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3299
- Antigen synonyms:CTRCT5|HSTF 4|ldis1|mHSF4|HSF 4|CTM|HSTF4
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: HSF4
Clonality: Monoclonal
Clone: 2B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human