Anti-HTT Mouse Monoclonal Antibody [clone: 1A12]
Catalog # ABNOH00003064-M13
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:huntingtin
- Antigen symbol:HTT
- Clonality:Monoclonal
- Clone:1A12
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:3064
- Antigen synonyms:HD|IT15
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: HTT
Clonality: Monoclonal
Clone: 1A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human