Order Entry
Poland
ContactUsLinkComponent
Anti-GSS Mouse Monoclonal Antibody [clone: 4B6]
Anti-GSS Mouse Monoclonal Antibody [clone: 4B6]
  ABNOH00002937-M03
 :  Abnova
undefined
Anti-GSS Mouse Monoclonal Antibody [clone: 4B6]
  ABNOH00002937-M03
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Glutathione synthetase
  • Antigen symbol:
    GSS
  • Clonality:
    Monoclonal
  • Clone:
    4B6
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    2937
  • Antigen synonyms:
    GSH synthetase|GSH-S|OTTHUMP00000030711|MGC14098|GSH S|GSHS|GSHB|GSS|Glutathione synthase
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: GSS
Clonality: Monoclonal
Clone: 4B6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human