Anti-GRIA3 Mouse Monoclonal Antibody [clone: 3D10]
ABNOH00002892-M04
: Abnova
- Antibody type:Primary
- Antigen name:Glutamate receptor 3
- Antigen symbol:GRIA3
- Clonality:Monoclonal
- Clone:3D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:2892
- Antigen synonyms:GluA3|GLUR3|glutamate receptor 3|AMPA-selective glutamate receptor 3|GLURC|glutamate receptor ionotropic|gluR-3|AMPA 3|glutamate receptor C|GLUR-K3|glutamate receptor subunit 3|GLUR-C|MRX94|dJ1171F9.1|RP11-349N19.3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: GRIA3
Clonality: Monoclonal
Clone: 3D10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human