Anti-FGF5 Mouse Polyclonal Antibody
Catalog # ABNOH00002250-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Fibroblast growth factor 5
- Antigen symbol:FGF5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2250
- Antigen synonyms:TCMGLY|HBGF-5|Smag-82
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: FGF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human