Anti-MECOM Mouse Monoclonal Antibody [clone: 6A9]
ABNOH00002122-M01
: Abnova
- Antibody type:Primary
- Antigen name:MDS1 and EVI1 complex locus
- Antigen symbol:MECOM
- Clonality:Monoclonal
- Clone:6A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:2122
- Antigen synonyms:AML1-EVI-1|MDS1|MDS1-EVI1|EVI1|PRDM3
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: MECOM
Clonality: Monoclonal
Clone: 6A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human