Anti-CYB5R3 Mouse Monoclonal Antibody [clone: 2A9]
Catalog # ABNOH00001727-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cytochrome b5 reductase 3
- Antigen symbol:CYB5R3
- Clonality:Monoclonal
- Clone:2A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:1727
- Antigen synonyms:0610016L08Rik|WU:AL591952.1-001|diaphorase 1|Dia-1|WU:Cyb5r3|Dia1|2500002N19Rik|WU:AL591952.1-002|diaphorase 1 (NADH)|NADH-cytochrome b5 reductase|DIA1|Cyb5r3|C85115|WU:AL591952.1-003
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CYB5R3
Clonality: Monoclonal
Clone: 2A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human