Order Entry
Poland
ContactUsLinkComponent
 
Anti-AKR1C2 Mouse Polyclonal Antibody
  ABNOH00001646-B02P
 :  Abnova
undefined
Anti-AKR1C2 Mouse Polyclonal Antibody
  ABNOH00001646-B02P
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Aldo-keto reductase family 1 member C2
  • Antigen symbol:
    AKR1C2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    1646
  • Antigen synonyms:
    TDD|DD|BABP|DDH2|AKR1C-pseudo|SRXY8|DD2|DD/BABP|HBAB|DD-2|MCDR2|HAKRD
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µg
  • Pk:
    50 µg

 

 

Type: Primary
Antigen: AKR1C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human