Anti-CSNK2A2 Mouse Monoclonal Antibody [clone: 1E8]
ABNOH00001459-M03
: Abnova
- Antibody type:Primary
- Antigen name:Casein Kinase 2, Alpha 2
- Antigen symbol:CSNK2A2
- Clonality:Monoclonal
- Clone:1E8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1459
- Antigen synonyms:CK2A2|CSNK2A1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CSNK2A2
Clonality: Monoclonal
Clone: 1E8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human