Anti-CSF2 Mouse Monoclonal Antibody [clone: 2G5]
ABNOH00001437-M08
: Abnova
- Antibody type:Primary
- Antigen name:Colony Stimulating Factor 2
- Antigen symbol:CSF2
- Clonality:Monoclonal
- Clone:2G5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1437
- Antigen synonyms:GMCSF|TUBB2|Csfgm|Gm-CSf|TUBB|MGI-IGM|CDCBM5
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: 2G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human