Order Entry
Poland
ContactUsLinkComponent
Anti-CSE1L Mouse Monoclonal Antibody [clone: 2C10]
Anti-CSE1L Mouse Monoclonal Antibody [clone: 2C10]
Catalog # ABNOH00001434-M03A
Supplier:  Abnova
undefined
Anti-CSE1L Mouse Monoclonal Antibody [clone: 2C10]
Catalog # ABNOH00001434-M03A
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Chromosome segregation 1-like
  • Antigen symbol:
    CSE1L
  • Clonality:
    Monoclonal
  • Clone:
    2C10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    1434
  • Antigen synonyms:
    XPO2|Exportin-2|CSE-1|CAS|XPO-2|CSE1L|Chromosome Segregation 1-like|Cellular Apoptosis Susceptibility Protein|CSE1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: CSE1L
Clonality: Monoclonal
Clone: 2C10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human