Anti-CREM Rabbit Polyclonal Antibody
100 µg
Catalog # ABNOH00001390-D01P
Supplier: Abnova
Jump to:
Product Details & Documents
Type: Primary
Antigen: CREM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Documents
TDS_ABNOH00001390-D01P.pdf
Datasheet
Specifications
- Size:100 µg
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Storage temperature:Cool
- Host:Rabbit
- Cross adsorption:No
- Shipping temperature:Ice
- Antigen name:cAMP responsive element modulator
- Cat. No.:ABNOH00001390-D01P
- Reactivity:Human
- Gene ID:1390
- Antigen synonyms:hCREM-2|CREM-2|ICER
- Conjugation:Unconjugated
- Antigen symbol:CREM



