Anti-TBCB Mouse Polyclonal Antibody
Catalog # ABNOH00001155-B02P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Tubulin Folding Cofactor B
- Antigen symbol:TBCB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1155
- Antigen synonyms:CKAPI|CKAP1|CG22
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: TBCB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human