Anti-CD33 Mouse Polyclonal Antibody
ABNOH00000945-B01P
: Abnova
- Antibody type:Primary
- Antigen name:CD33 molecule
- Antigen symbol:CD33
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:945
- Antigen synonyms:SIGLEC-3|SIGLEC3|CD33|Siglec-3|Myeloid cell surface antigen CD33|Sialic acid-binding Ig-like lectin 3|p67
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: CD33
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human