Anti-CD8B Mouse Polyclonal Antibody
50 µg
ABNOH00000926-B01P
: Abnova
Type: Primary
Antigen: CD8B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
- Size:50 µg
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Storage temperature:Cool
- Host:Mouse
- Cross adsorption:No
- Shipping temperature:Ice
- Antigen name:CD8b molecule
- Cat. No.:ABNOH00000926-B01P
- Reactivity:Human
- Gene ID:926
- Antigen synonyms:LY3|LYT3|OX-8 membrane antigen|CD8 antigen beta chain|LEU2|T-cell surface glycoprotein CD8 beta chain|P37|Cd8b1|CD8 antigen 37 kDa chain|CD8 antigen beta-chain
- Conjugation:Unconjugated
- Antigen symbol:CD8B
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



