Anti-CCND2 Mouse Monoclonal Antibody [clone: 3B10]
Catalog # ABNOH00000894-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Cyclin D2
- Antigen symbol:CCND2
- Clonality:Monoclonal
- Clone:3B10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:894
- Antigen synonyms:G1/S-specific cyclin-D2|MPPH3|G1/S-specific cyclin D2|MGC102758|KIAK0002
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CCND2
Clonality: Monoclonal
Clone: 3B10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human