Order Entry
Poland
ContactUsLinkComponent
Anti-TSPO Mouse Polyclonal Antibody
Anti-TSPO Mouse Polyclonal Antibody
Catalog # ABNOH00000706-A01
Supplier:  Abnova
Anti-TSPO Mouse Polyclonal Antibody
Catalog # ABNOH00000706-A01
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Translocator Protein (18kDa)
  • Antigen symbol:
    TSPO
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Gene ID:
    706
  • Antigen synonyms:
    peripheral-type benzodiazepine receptor|MBR|DBI|benzodiazapine receptor (peripheral)|peripheral benzodiazepine receptor|IBP|OTTHUMP00000028891|pk18 |translocator protein (18kDa) |TSPO|PTBR|BZRP|PKBS|mitochondrial benzodiazepine receptor|benzodiazepine peripheral binding site|mDRC|PBR
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    50 µl
  • Pk:
    50 µl

Specifications

About this item

Type: Primary
Antigen: TSPO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human