Anti-BMP5 Mouse Monoclonal Antibody [clone: 4A9]
ABNOH00000653-M08
: Abnova
- Antibody type:Primary
- Antigen name:Bone morphogenetic Protein 5
- Antigen symbol:BMP5
- Clonality:Monoclonal
- Clone:4A9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:653
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: BMP5
Clonality: Monoclonal
Clone: 4A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human