Anti-BAK Mouse Monoclonal Antibody [clone: 1E4]
ABNOH00000578-M01A
: Abnova
- Antibody type:Primary
- Antigen name:BCL2-Antagonist/Killer 1
- Antigen symbol:BAK
- Clonality:Monoclonal
- Clone:1E4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:578
- Antigen synonyms:bcl-2 homologous antagonist/killer|bcl-2-like protein 7|BCL2-like 7 protein|pro-apoptotic protein BAK|BAK1|bcl2-L-7|apoptosis regulator BAK
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: BAK
Clonality: Monoclonal
Clone: 1E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human