Anti-ATP1B3 Mouse Polyclonal Antibody
ABNOH00000483-B01P
: Abnova
- Antibody type:Primary
- Antigen name:ATPase, Na+/K+ transporting, beta 3 polypeptide
- Antigen symbol:ATP1B3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:483
- Antigen synonyms:ATPB-3|CD298
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Type: Primary
Antigen: ATP1B3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human