Anti-ASCL1 Mouse Monoclonal Antibody [clone: 2D9]
ABNOH00000429-M02
: Abnova
- Antibody type:Primary
- Antigen name:Achaete-scute Homolog 1
- Antigen symbol:ASCL1
- Clonality:Monoclonal
- Clone:2D9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:429
- Antigen synonyms:ASH-1|ASH1|Mash1|BHLHA46|HASH1|Mash-1|hASH1|bHLHa46|Class A basic helix-loop-helix protein 46|Achaete-scute homolog 1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Type: Primary
Antigen: ASCL1
Clonality: Monoclonal
Clone: 2D9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human