Anti-KLK3 Mouse Monoclonal Antibody [clone: 1B1]
Catalog # ABNOH00000354-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Kallikrein-related peptidase 3
- Antigen symbol:KLK3
- Clonality:Monoclonal
- Clone:1B1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:354
- Antigen synonyms:PSA|APS|hK3|KLK2A1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: KLK3
Clonality: Monoclonal
Clone: 1B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human