Anti-APOC2 Mouse Monoclonal Antibody [clone: 3E4]
Catalog # ABNOH00000344-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Apolipoprotein C2
- Antigen symbol:APOC2
- Clonality:Monoclonal
- Clone:3E4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:344
- Antigen synonyms:MGC75082|APO-CII|APOC-II
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: APOC2
Clonality: Monoclonal
Clone: 3E4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human