Anti-AHCY Mouse Monoclonal Antibody [clone: M1]
ABNOH00000191-M08A
: Abnova
- Antibody type:Primary
- Antigen name:Adenosylhomocysteinase
- Antigen symbol:AHCY
- Clonality:Monoclonal
- Clone:M1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:191
- Antigen synonyms:adoHcyase|SAHH
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Type: Primary
Antigen: AHCY
Clonality: Monoclonal
Clone: M1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human