Order Entry
Poland
ContactUsLinkComponent
Anti-ABCA1 Mouse Monoclonal Antibody [clone: 1H4]
Anti-ABCA1 Mouse Monoclonal Antibody [clone: 1H4]
Catalog # ABNOH00000019-M01A
Supplier:  Abnova
Anti-ABCA1 Mouse Monoclonal Antibody [clone: 1H4]
Catalog # ABNOH00000019-M01A
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    ATP Binding Cassette, sub-family A, member 1
  • Antigen symbol:
    ABCA1
  • Clonality:
    Monoclonal
  • Clone:
    1H4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    19
  • Antigen synonyms:
    ABC-1|ATP-binding cassette transporter 1|CERP|HDLDT1|TGD|ABC1|membrane-bound|ATP-binding cassette transporter A1|cholesterol efflux regulatory protein
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    200 µl
  • Pk:
    200 µl

Specifications

About this item

Type: Primary
Antigen: ABCA1
Clonality: Monoclonal
Clone: 1H4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human