Anti-MAP2K1 Mouse Monoclonal Antibody [clone: OTI1A10]
Catalog # ORIGTA600341
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase kinase 1
- Antigen symbol:MAP2K1
- Clonality:Monoclonal
- Clone:OTI1A10
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Stored in PBS (pH 7,4) containing 0,05% sodium azide and up to 5% trehalose
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Recommended Dilutions:
Type: Primary
Antigen: MAP2K1 (mitogen-activated protein kinase kinase 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human