Anti-PSMB9 Mouse Monoclonal Antibody [clone: OTI2B6]
ORIGTA504392
: OriGene
- Antibody type:Primary
- Antigen name:Proteasome (prosome, macropain) subunit, Beta type 9
- Antigen symbol:PSMB9
- Clonality:Monoclonal
- Clone:OTI2B6
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:21 to 219
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:23.1 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150
Type: Primary
Antigen: PSMB9 (proteasome (prosome, macropain) subunit, beta type, 9)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: