Anti-FMR1 Mouse Monoclonal Antibody [clone: OTI2C3]
numer katalogowy ORIGTA504293
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Fragile X mental retardation 1
- Antigen symbol:FMR1
- Clonality:Monoclonal
- Clone:OTI2C3
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:26 to 279
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:71 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:200-1000; Immunochemistry: 1:150
Type: Primary
Antigen: FMR1 (fragile X mental retardation 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: