Anti-STAT5A Mouse Monoclonal Antibody [clone: OTI4E7]
Catalog # ORIGTA502738
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Signal transducer and activator of transcription 5A
- Antigen symbol:STAT5A
- Clonality:Monoclonal
- Clone:OTI4E7
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:90.5 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500-2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: STAT5A (signal transducer and activator of transcription 5A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: