Anti-PDLIM2 Mouse Monoclonal Antibody [clone: OTI11A12]
ORIGTA502731
: OriGene
- Antibody type:Primary
- Antigen name:PDZ And LIM Domain 2
- Antigen symbol:PDLIM2
- Clonality:Monoclonal
- Clone:OTI11A12
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:37.3 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:100; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: PDLIM2 (PDZ and LIM domain 2 (mystique))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human