Anti-VWA5A Mouse Monoclonal Antibody [clone: OTI3B4]
ORIGTA501654
: OriGene
- Antibody type:Primary
- Antigen name:von Willebrand factor A domain containing 5A
- Antigen symbol:VWA5A
- Clonality:Monoclonal
- Clone:OTI3B4
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:86.3 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: VWA5A (von Willebrand factor A domain containing 5A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human