Anti-ID2 Mouse Monoclonal Antibody [clone: OTI10C3]
Catalog # ORIGTA500183
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Inhibitor of DNA binding 2, dominant negative helix-loop-helix Protein
- Antigen symbol:ID2
- Clonality:Monoclonal
- Clone:OTI10C3
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Human
- Western blot:Yes
- Epitope:Full length
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:14.7 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Pk:100 µl
Specifications
About this item
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:50
Type: Primary
Antigen: ID2 (inhibitor of DNA binding 2, dominant negative helix-loop-helix protein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human