Order Entry
Poland
ContactUsLinkComponent
Mouse IgG2b Isotype Control [clone: 1D4]
Mouse IgG2b Isotype Control [clone: 1D4]
Catalog # ORIGTA310153
Supplier:  OriGene
Mouse IgG2b Isotype Control [clone: 1D4]
Catalog # ORIGTA310153
Supplier:  OriGene

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    1D4
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS, pH 7,4 with 0,05% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µl

Specifications

About this item

  • Recommended Dilutions:

Type: Control
Antigen: GAPDH (glyceraldehyde-3-phosphate dehydrogenase)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: