Anti-FANCD2 Mouse Monoclonal Antibody [clone: FI-17]
ORIGTA309656
: OriGene
- Antibody type:Primary
- Antigen name:Fanconi anemia, complementation group D2
- Antigen symbol:FANCD2
- Clonality:Monoclonal
- Clone:FI-17
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Format:Ascites
- Form:Liquid
- Storage buffer:0,1% Sodium Azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 ml
Recommended Dilutions:
Type: Primary
Antigen: FANCD2 (Fanconi anemia, complementation group D2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human