Anti-POLR3F Rabbit Polyclonal Antibody
Catalog # ORIGTA306617
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Polymerase (RNA) III (DNA directed) polypeptide F (39kDa)
- Antigen symbol:POLR3F
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Storage buffer:PBS containing 0,02% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography via peptide column
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml; Immunochemistry: 2.5 ?g/ml; Immunofluorescence: 20 ?g/ml
Type: Primary
Antigen: POLR3F (polymerase (RNA) III (DNA directed) polypeptide F, 39 kDa)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat